Invitrogen
Manufacturer Code:PA533447
Catalog # PIPA533447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from N-terminal extracellular domain of human CRF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR-1; CRH receptor 1; CRH-R 1; CRH-R-1; CRH-R1; CRHR1 CRF-R-1 CRFR CRHR1f Crf receptor 1 CRH-R1 CRF-R CRF1 CRFR-1 Crf receptor type 1 Crf receptor type 1 Crf type 1 CRF-R1 Crf1 receptor CRFR1 CRH-R-1 CRH-R1h CRHR CRHR1L; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998, (Rat) P35353
Entrez Gene ID: (Human) 1394, (Rat) 58959
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.