Invitrogen
Manufacturer Code:PA527121
Catalog # PIPA527121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 53 and 147 of Human CRFR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF 1 receptor; CRF-R CRF-R-1 CRF-R1 CRF1 CRFR-1 CRFR1 CRH-R-1 CRH-R1 CRH-R1h CRHR CRHR1L CRHR1f corticotropin releasing hormone receptor variant 1e corticotropin releasing hormone receptor variant 1g corticotropin-releasing factor receptor 1 corticotropin-releasing factor type 1 receptor corticotropin-releasing hormone receptor 1 seven transmembrane; CRF-R-1; CRF-R1; CRF-R1alpha; CRFR-1; CRH receptor 1; CRH receptor subtype-1 isoform-3; CRH-R-1; CRH-R1; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998, (Mouse) P35347
Entrez Gene ID: (Human) 1394, (Mouse) 12921
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.