Invitrogen
Manufacturer Code:PA518801
Catalog # PIPA518801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NEEKKSKVHYH) corresponding to the internal amino acids of CRHR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF 1 receptor; CRF-R; CRF-R-1; CRF-R1; CRF-R1alpha; CRF1; CRFR-1; CRH receptor 1; CRH receptor subtype-1 isoform-3; CRH-R 1; CRH-R-1; CRH-R1; CRHR1 CRF-R corticotropin releasing hormone receptor 1 CRF1 CRFR1 CRH-R1h CRHR CRHR1f corticotropin releasing hormone receptor variant 1h seven transmembrane helix receptor; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998, (Rat) P35353, (Mouse) P35347
Entrez Gene ID: (Human) 1394, (Rat) 58959, (Mouse) 12921
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.