Abnova
Manufacturer Code:PAB16403
Catalog # 89-118-275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human CRHR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CRHR1.
For Research Use Only
Protein Aliases: Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF-R; CRF-R-1; CRF1; CRFR1; CRH receptor 1; CRH-R-1; CRH-R1h; CRHR; CRHR1f; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998
Entrez Gene ID: (Human) 1394
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.