Abnova
Manufacturer Code:H00001394K
Catalog # 89-205-491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CRHR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CRHR1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF-R CRF1 CRFR1 CRH-R1h CRHR CRHR1f; CRF-R-1; CRH receptor 1; CRH-R-1; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998
Entrez Gene ID: (Human) 1394
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.