Invitrogen
Manufacturer Code:PA548025
Catalog # PIPA548025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse CRHBP Tyr25-Leu322 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticotropin-releasing factor-binding protein; Corticotropin-releasing hormone-binding protein; Corticotropin-releasing hormone-binding protein CRH-BP Corticotropin-releasing factor-binding protein CRF-binding protein CRF-BP CRHBP CRFBP; CRF-binding protein; CRF-BP; CRH-BP
Gene Aliases: CRH-BP; Crhbp
UniProt ID: (Mouse) Q60571
Entrez Gene ID: (Mouse) 12919
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.