R&D Systems
Manufacturer Code:BAF2796
Catalog # BAF2796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CRHBP Tyr25-Leu322 Accession # P24387.2 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: corticotropin releasing hormone binding protein corticotropin-releasing factor-binding protein Corticotropin-releasing hormone-binding protein CRFBP CRFBPcorticotropin releasing hormone-binding protein CRF-BPCRF-binding protein CRH-BP; corticotropin releasing hormone-binding protein; Corticotropin-releasing factor-binding protein; Corticotropin-releasing hormone-binding protein; CRF-binding protein; CRF-BP; CRH-BP
Gene Aliases: CRF-BP; CRFBP; CRHBP
UniProt ID: (Human) P24387
Entrez Gene ID: (Human) 1393
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.