Invitrogen
Manufacturer Code:PA5116529
Catalog # PIPA5116529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 25-194 of human CRH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticoliberin; Corticoliberin corticotropin releasing factor corticotropin releasing hormone Corticotropin-releasing factor Corticotropin-releasing hormone CRF Crh CRH1 Gm1347; corticotropin releasing factor; Corticotropin-releasing factor; Corticotropin-releasing hormone; CRF
Gene Aliases: CRF; CRH; CRH1; Gm1347
UniProt ID: (Human) P06850, (Rat) P01143, (Mouse) Q8CIT0
Entrez Gene ID: (Human) 1392, (Rat) 81648, (Mouse) 12918
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.