Invitrogen
Manufacturer Code:PA5113738
Catalog # PIPA5113738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Human CRH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticoliberin; Corticoliberin corticotrophin releasing hormone receptor 2 corticotropin releasing factor corticotropin releasing factor receptor type 2 beta corticotropin releasing hormone corticotropin releasing hormone receptor 2 Corticotropin-releasing factor corticotropin-releasing factor 2 Corticotropin-releasing factor receptor 2 corticotropin-releasing factor receptor subtype 2 Corticotropin-releasing hormone Corticotropin-releasing hormone receptor 2 CRF CRF 2 receptor CRF2 CRF2 receptor beta isoform CRF2R CRF-R 2 CRFR2 CRF-R2 CRF-R-2 CRFR-2 CRFR2alpha CRFR2beta CRF-RB CRH CRH receptor 2 variant B CRH1 CRH2R CRH-R 2 CRHR2 CRH-R2 CRH-R-2 Gm1347 HM-CRF; Corticotropin-releasing factor; Corticotropin-releasing hormone; CRF
Gene Aliases: CRF; CRH; CRH1
UniProt ID: (Human) P06850
Entrez Gene ID: (Human) 1392
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.