Invitrogen
Manufacturer Code:PA5111543
Catalog # PIPA5111543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human CRF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticoliberin; Corticoliberin Corticotropin-releasing factor Corticotropin-releasing factor receptor 2 Corticotropin-releasing hormone Corticotropin-releasing hormone receptor 2 CRF CRF2 CRF2R Crfr2 CRF-R2 CRF-R-2 CRFR2alpha CRFR2beta CRF-RB CRH CRH1 CRH2R Crhr2 CRH-R-2 Gm1347 HM-CRF; Corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; Corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF-R-2; CRF2 receptor, beta isoform; CRH receptor 2 variant B; CRH-R-2; CRH-R2
Gene Aliases: CRF; CRF-RB; CRF2; CRF2R; CRFR2; CRH; CRH1; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) P06850, (Human) Q13324
Entrez Gene ID: (Human) 1392, (Human) 1395
Molecular Function:
peptide hormone
signaling molecule
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.