Invitrogen
Manufacturer Code:PA137498
Catalog # 01-673-319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from C-terminus of human CRF protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze,do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticoliberin; Corticotropin Releasing Factor; Corticotropin-releasing factor; Corticotropin-releasing hormone; CRF
Gene Aliases: CRF; CRH; CRH1
UniProt ID: (Human) P06850
Entrez Gene ID: (Human) 1392
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.