Proteintech
Manufacturer Code:124391AP150UL
Catalog # 50-172-7981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CREST Fusion Protein Ag3119 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-responsive transactivator; Calcium-responsive transcription coactivator; CREST KIAA0693 LP2261 SS18 like protein 1 SS18L1 SYT homolog 1; SS18-like protein 1; synovial sarcoma translocation gene on chromosome 18-like 1; SYT homolog 1
Gene Aliases: A230053O16Rik; CREST; KIAA0693; LP2261; SS18L1
UniProt ID: (Human) Q8NE69, (Mouse) Q8BW22, (Rat) Q91XJ0
Entrez Gene ID: (Human) 26039, (Mouse) 269397, (Rat) 192352
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.