Invitrogen
Manufacturer Code:PA568552
Catalog # PIPA568552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human CREBRF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adult retina protein; C5orf41; CREB3 regulatory factor; LRF; Luman recruitment factor; luman-recruiting factor; luman/CREB3 recruitment factor; UPF0474 protein C5orf41; UPF0474 protein C5orf41 homolog
Gene Aliases: A930001N09Rik; C4H5orf41; C5orf41; CREBRF; gb:bc142922; LRF; RGD1310862
UniProt ID: (Human) Q8IUR6, (Mouse) Q8CDG5
Entrez Gene ID: (Horse) 100059002, (Dog) 612888, (Pig) 100523409, (Human) 153222, (Rabbit) 100341765, (Rat) 303016, (Mouse) 77128, (Zebrafish) 100149876
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.