Novus Biologicals
Manufacturer Code:NBP179997
Catalog # NBP179997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LOC153222. Peptide sequence DDEDHDEGFGSEHELSENEEEEEEEEDYEDDKDDDISDTFSEPGYEND. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adult retina protein; adult retina protein C5orf41 chromosome 5 open reading frame 41 CREB3 regulatory factor CREBRF DKFZp313F2319 DKFZp686G2059 LRF luman-recruiting factor; CREB3 regulatory factor; LRF; Luman recruitment factor; luman-recruiting factor; luman/CREB3 recruitment factor; UPF0474 protein C5orf41
Gene Aliases: C5orf41; CREBRF; LRF
UniProt ID: (Human) Q8IUR6
Entrez Gene ID: (Human) 153222
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.