Abnova
Manufacturer Code:PAB25857
Catalog # 89-127-486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S129 of human CREB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CREB1.
For Research Use Only
Protein Aliases: active transcription factor CREB; cAMP-response element-binding protein-1; CREB; CREB-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; Cyclic AMP-responsive element-binding protein 1; MGC9284; transactivator protein
Gene Aliases: CREB; CREB-1; CREB1
UniProt ID: (Human) P16220
Entrez Gene ID: (Human) 1385
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.