Novus Biologicals
Manufacturer Code:NBP292783100UL
Catalog # NB027432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human CRCP (NP_001035737.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcitonin gene-related peptide-receptor component protein; Calcitonin gene-related peptide-receptor component protein CGRP receptor component CGRPRCP CGRP-RCPRNA polymerase III subunit C9 CGRP-receptor component protein DNA-directed RNA polymerase III subunit RPC9 hsC17 RCP9 RCPMGC111194; CGRP-RCP; CGRP-receptor component protein; DNA-directed RNA polymerase III subunit RPC9; RNA polymerase III subunit C9
Gene Aliases: CGRP-RCP; CGRPRCP; CRCP; RCP; RCP9
UniProt ID: (Human) O75575
Entrez Gene ID: (Human) 27297
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.