Proteintech
Manufacturer Code:151701AP150UL
Catalog # 50-173-0038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CRAT Fusion Protein Ag7422 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CACP Carnitine acetylase carnitine acetyltransferase Carnitine O acetyltransferase CAT CAT1 CRAT; Carnitine acetylase; Carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT
Gene Aliases: AW107812; CARAT; CAT1; CRAT
UniProt ID: (Human) P43155, (Rat) Q704S8, (Mouse) P47934
Entrez Gene ID: (Human) 1384, (Rat) 311849, (Mouse) 12908
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.