Novus Biologicals
Manufacturer Code:NBP245868
Catalog # NBP245868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F1 |
Immunogen | Full length human recombinant protein of human SLAMF7 (NP_067004) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 19A CD2 subset 1 CD2-like receptor activating cytotoxic cells CD319 CD319 antigen CRACCCD2-like receptor-activating cytotoxic cells CS119A24 protein Membrane protein FOAP-12 Novel Ly9 novel LY9 (lymphocyte antigen 9) like protein Protein 19A SLAM family member 7; 19A24 protein; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; Membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; Protein 19A; SLAM family member 7
Gene Aliases: 19A; CD319; CRACC; CS1; SLAMF7; UNQ576/PRO1138
UniProt ID: (Human) Q9NQ25
Entrez Gene ID: (Human) 57823
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
membrane-bound signaling molecule
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.