Proteintech
Manufacturer Code:664681IG150UL
Catalog # 50-173-6892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A5F3 |
Immunogen | CRABP2 Fusion Protein Ag0309 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cellular lipid-binding protein; cellular retinoic acid binding protein II; Cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; CRABP II CRABP2 RBP6; CRABP-II
Gene Aliases: AI893628; Crabp-2; CRABP-II; CRABP2; CrabpII; RBP6
UniProt ID: (Human) P29373, (Mouse) P22935, (Rat) P51673
Entrez Gene ID: (Human) 1382, (Pig) 100155151, (Mouse) 12904, (Rat) 29563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.