Invitrogen
Manufacturer Code:PA525337
Catalog # PIPA525337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 90-118 amino acids from the C-terminal region of human CRABP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; CRABP CRABP-I CRABPI RBP5 cellular retinoic acid-binding protein 1 cellular retinoic acid-binding protein I; CRABP-I
Gene Aliases: AI326249; CRABP; Crabp-1; CRABP-I; CRABP1; CRABPI; Rbp-5; RBP5
UniProt ID: (Human) P29762, (Mouse) P62965
Entrez Gene ID: (Human) 1381, (Mouse) 12903
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.