Novus Biologicals
Manufacturer Code:NBP304041
Catalog # NB093723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human CRABP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cellular retinoic acid binding protein 1 CRABP CRABPI CRABP-Icellular retinoic acid-binding protein 1 RBP5Cellular retinoic acid-binding protein I; Cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; CRABP-I
Gene Aliases: CRABP; CRABP-I; CRABP1; CRABPI; RBP5
UniProt ID: (Human) P29762
Entrez Gene ID: (Human) 1381
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.