Abnova
Manufacturer Code:MAB12161
Catalog # 89-110-302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP186 |
Immunogen | A synthetic peptide corresponding to C-terminus region of human CR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit monoclonal antibody raised against synthetic peptide of human CR2.
For Research Use Only
Protein Aliases: C3DR; CD21; Complement C3d receptor; complement component (3d/Epstein Barr virus) receptor 2; Complement receptor type 2; Cr2; EBV receptor; Epstein-Barr virus receptor; SLEB9
Gene Aliases: C3DR; CD21; CR; CR2; CVID7; SLEB9
UniProt ID: (Human) P20023
Entrez Gene ID: (Human) 1380
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.