Proteintech
Manufacturer Code:125481AP150UL
Catalog # 50-172-8056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPVL Fusion Protein Ag3398 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carboxypeptidase WUG; Carboxypeptidase, vitellogenic-like; CP-Mac carboxypeptidase; CPVL HVLP UNQ197/PRO223 VCP like protein VLP; Probable serine carboxypeptidase CPVL; VCP-like protein; Vitellogenic carboxypeptidase-like protein
Gene Aliases: 4933436L16Rik; CPVL; HVLP; PSEC0124; UNQ197/PRO223; VLP
UniProt ID: (Human) Q9H3G5, (Rat) Q4QR71, (Mouse) Q9D3S9
Entrez Gene ID: (Human) 54504, (Rat) 502774, (Mouse) 71287
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.