Invitrogen
Manufacturer Code:PA527999
Catalog # PIPA527999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 9 and 359 of Human CPT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 2, mitochondrial; Carnitine palmitoyltransferase II; CPT II; CPT1 CPTASE IIAE4 CPT II carnitine O-palmitoyltransferase 2 mitochondrial carnitine palmitoyltransferase II; testicular secretory protein Li 13
Gene Aliases: CPT1; CPT2; CPTASE; IIAE4
UniProt ID: (Human) P23786
Entrez Gene ID: (Human) 1376
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.