Novus Biologicals
Manufacturer Code:NBP146137
Catalog # NBP146137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TDSTVTVQKLNFE corresponding to internal region according to NP_000089.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: carnitine O-palmitoyltransferase 2 mitochondrial carnitine palmitoyltransferase 2 Carnitine palmitoyltransferase IIEC 2.3.1.21 CPT II CPT1 CPTASE; Carnitine O-palmitoyltransferase 2, mitochondrial; Carnitine palmitoyltransferase II; CPT II; testicular secretory protein Li 13
Gene Aliases: CPT1; CPT2; CPTASE; IIAE4
UniProt ID: (Human) P23786
Entrez Gene ID: (Human) 1376
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.