Proteintech
Manufacturer Code:265551AP150UL
Catalog # 50-173-5375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPT2 Fusion Protein Ag24897 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 2, mitochondrial; Carnitine palmitoyltransferase II; CPT II; CPT II CPT1 CPT2 CPTASE; testicular secretory protein Li 13
Gene Aliases: AI323697; Cpt-2; CPT1; CPT2; CPTASE; CPTII; IIAE4
UniProt ID: (Human) P23786, (Mouse) P52825, (Rat) P18886
Entrez Gene ID: (Human) 1376, (Mouse) 12896, (Rat) 25413
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.