Invitrogen
Manufacturer Code:PA589482
Catalog # PIPA589482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 270-400 of human CPT1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 1, brain isoform; Carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; Carnitine palmitoyltransferase 1C; CPT I-C; CPT IC; CPT1-B; CPTI-B
Gene Aliases: 6530437J22Rik; 9630004I06Rik; CPT IC; CPT1-B; Cpt1c; CPTI-B
UniProt ID: (Rat) F1LN46, (Mouse) Q8BGD5
Entrez Gene ID: (Rat) 308579, (Mouse) 78070
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.