Invitrogen
Manufacturer Code:PA584607
Catalog # PIPA584607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CPT1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 1, brain isoform; Carnitine O-palmitoyltransferase I, brain isoform; Carnitine palmitoyltransferase 1C; carnitine palmitoyltransferase I related C; CPT IC; CPT1-B; CPT1P CPTIC FLJ23809; CPTI-B
Gene Aliases: CATL1; CPT1-B; CPT1C; CPT1P; CPTI-B; CPTIC; SPG73
UniProt ID: (Human) Q8TCG5
Entrez Gene ID: (Human) 126129
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.