Novus Biologicals
Manufacturer Code:NBP29213320UL
Catalog # NB026159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 270-400 of human CPT1C (NP_001129524.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carnitine O-palmitoyltransferase 1 brain isoform Carnitine O-palmitoyltransferase I brain isoform carnitine palmitoyltransferase 1CCPT1-B carnitine palmitoyltransferase I related C CATL1 CPT IC CPT1P CPTI-B CPTIC EC 2.3.1.21 FLJ23809; Carnitine O-palmitoyltransferase 1, brain isoform; Carnitine O-palmitoyltransferase I, brain isoform; Carnitine palmitoyltransferase 1C; carnitine palmitoyltransferase I related C; CPT IC; CPT1-B; CPTI-B
Gene Aliases: CATL1; CPT1-B; CPT1C; CPT1P; CPTI-B; CPTIC; SPG73
UniProt ID: (Human) Q8TCG5
Entrez Gene ID: (Human) 126129
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.