Invitrogen
Manufacturer Code:PA569410
Catalog # PIPA569410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human CPT1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; Carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; Carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase I; Carnitine palmitoyltransferase I-like protein; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; CPT I; CPT-Ibeta 1; CPT-Ibeta 3; CPT1-M; CPT1B carnitine palmitoyltransferase I muscle carnitine palmitoyltransferase 1B (muscle) CPT1-M M-CPT1 KIAA1670 CPTI muscle carnitine O-palmitoyltransferase I mitochondrial muscle isoform carnitine palmitoyltransferase 1A (muscle) carnitine palmitoyltransferase 1B carnitine palmitoyltransferase 1B (muscle) CPT1M FLJ55729 FLJ58750 MCCPT1 MCPT1 carnitine palmitoyltransferase I-like; CPTI-M; muscle-type carnitine palmitoyltransferase I
Gene Aliases: CPT-IB; Cpt1; CPT1-M; cpt1al; CPT1B; CPT1M; CPTI; CPTI-M; CPTIB; KIAA1670; M-CPT1; M-CPTI; MCCPT1; MCPT1; zgc:103709
UniProt ID: (Human) Q92523, (Bovine) Q58DK1, (Mouse) Q924X2, (Rat) Q63704
Entrez Gene ID: (Rabbit) 100353138, (Human) 1375, (Horse) 100055438, (Sheep) 443193, (Dog) 481184, (Bovine) 509459, (Mouse) 12895, (Guinea pig) 100717974, (Rat) 25756, (Zebrafish) 449677
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.