Novus Biologicals
Manufacturer Code:NB1001057
Catalog # NB1001057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DIADLFQVPKAYS corresponding to C-Terminus according to NP_689452.1 NP_689451.1 NP_004368.1 NP_001138609.1 NP_001138607.1 NP_001138606.1 NP_001138608.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: carnitine O-palmitoyltransferase 1 muscle isoform carnitine O-palmitoyltransferase I mitochondrial muscle isoform Carnitine O-palmitoyltransferase I muscle isoform Carnitine palmitoyltransferase 1B carnitine palmitoyltransferase 1B (muscle) Carnitine palmitoyltransferase I-like protein CPT I CPT1M CPT1-MMCCPT1 CPTI CPTI-M EC 2.3.1 EC 2.3.1.21 FLJ55729 FLJ58750 KIAA1670 MCPT1 M-CPT1; Carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; Carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); Carnitine palmitoyltransferase I-like protein; CPT I; CPT1-M
Gene Aliases: CPT1-M; CPT1B; CPT1M; CPTI; CPTI-M; KIAA1670; M-CPT1; MCCPT1; MCPT1
UniProt ID: (Human) Q92523
Entrez Gene ID: (Human) 1375
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.