Invitrogen
Manufacturer Code:PA569347
Catalog # PIPA569347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human CPT1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carnitine O-palmitoyltransferase; Carnitine O-palmitoyltransferase 1, liver isoform; Carnitine O-palmitoyltransferase I, liver isoform; Carnitine palmitoyltransferase 1 alpha, liver isoform; carnitine palmitoyltransferase 1, liver; Carnitine palmitoyltransferase 1A; carnitine palmitoyltransferase 1A (liver); carnitine palmitoyltransferase 1a, liver; carnitine palmitoyltransferase I, liver; CPT I; CPT1-L; CPT1A carnitine palmitoyltransferase 1A (liver) CPT1 CPT1-L L-CPT1 carnitine palmitoyltransferase 1A carnitine palmitoyltransferase I carnitine palmitoyltransferase I liver CPT-1; CPTI-L; L-CPT I; mitochondrial carnitine palmitoyltransferase 1A
Gene Aliases: C730027G07; Cpt-1; CPT-Ia; CPT1; CPT1-L; CPT1A; CPTI; L-CPT1
UniProt ID: (Human) P50416, (Horse) Q68Y62, (Mouse) P97742, (Rat) P32198
Entrez Gene ID: (Dog) 403583, (Bovine) 506812, (Sheep) 443434, (Human) 1374, (Horse) 100009716, (Mouse) 12894, (Guinea pig) 100728420, (Rat) 25757
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.