Invitrogen
Manufacturer Code:PA518578
Catalog # PIPA518578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DPAQTVEQRLKLFK) corresponding to the internal amino acids of CPT1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnitine O-palmitoyltransferase 1, liver isoform; Carnitine O-palmitoyltransferase I, liver isoform; Carnitine palmitoyltransferase 1A; carnitine palmitoyltransferase 1A (liver); carnitine palmitoyltransferase I, liver; CPT I; CPT1-L; CPT1A carnitine palmitoyltransferase 1A (liver) CPT1 CPT1-L L-CPT1 carnitine palmitoyltransferase 1A carnitine palmitoyltransferase I carnitine palmitoyltransferase I liver CPT-1; CPTI-L
Gene Aliases: CPT1; CPT1-L; CPT1A; L-CPT1
UniProt ID: (Human) P50416
Entrez Gene ID: (Human) 1374
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.