Invitrogen
Manufacturer Code:PA541830
Catalog # PIPA541830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CPSF6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFIm68; Cleavage and polyadenylation specific factor 6 CFIM CFIM68 HPBRII-4 HPBRII-7; cleavage and polyadenylation specific factor 6, 68 kD subunit; cleavage and polyadenylation specific factor 6, 68kDa; Cleavage and polyadenylation specificity factor 68 kDa subunit; Cleavage and polyadenylation specificity factor subunit 6; Cleavage factor Im complex 68 kDa subunit; CPSF 68 kDa subunit; pre-mRNA cleavage factor I, 68kD subunit; pre-mRNA cleavage factor Im (68kD); Pre-mRNA cleavage factor Im 68 kDa subunit; Protein HPBRII-4/7
Gene Aliases: 4733401N12Rik; AI256641; CFIM; CFIM68; CPSF6; HPBRII-4; HPBRII-7; wu:fa22f12; zgc:85819
UniProt ID: (Human) Q16630, (Bovine) Q0P5D2, (Mouse) Q6NVF9, (Zebrafish) Q6NWC6
Entrez Gene ID: (Human) 11052, (Bovine) 768231, (Dog) 474441, (Rabbit) 100340648, (Horse) 100059688, (Mouse) 432508, (Guinea pig) 100732506, (Rat) 299811, (Zebrafish) 327069
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.