Invitrogen
Manufacturer Code:PA561877
Catalog # PIPA561877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CPSF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 4, 30kDa; Cleavage and polyadenylation specificity factor 30 kDa subunit; Cleavage and polyadenylation specificity factor subunit 4; CPSF 30 kDa subunit; CPSF30 NAR NEB1 CPSF 30 kDa subunit NS1 effector domain-binding protein 1 cleavage and polyadenylation specificity factor 30 kDa subunit cleavage and polyadenylation specificity factor subunit 4 cleavage-polyadenylation specificity factor 30kD neb-1 no arches homolog no; Neb-1; No arches homolog; no arches-like zinc finger protein; NS1 effector domain-binding protein 1
Gene Aliases: CPSF30; CPSF4; NAR; NEB-1; NEB1
UniProt ID: (Human) O95639
Entrez Gene ID: (Human) 10898
Molecular Function:
RNA binding protein
endoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.