Invitrogen
Manufacturer Code:PA557005
Catalog # PIPA557005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CPSF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; Cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF 73 kDa subunit; CPSF CPSF-73 CPSF73 YSH1 CPSF 73 kDa subunit cleavage and polyadenylation specificity factor 3 cleavage and polyadenylation specificity factor 73 kDa subunit cleavage and polyadenylation specificity factor subunit 3 mRNA 3&prime-end-processing endonuclease CPSF-73; mRNA 3'-end-processing endonuclease CPSF-73
Gene Aliases: CPSF-73; CPSF3; CPSF73
UniProt ID: (Human) Q9UKF6, (Mouse) Q9QXK7
Entrez Gene ID: (Human) 51692, (Mouse) 54451, (Rat) 298916
Molecular Function:
RNA binding protein
endoribonuclease
mRNA polyadenylation factor
mRNA processing factor
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.