Invitrogen
Manufacturer Code:PA531004
Catalog # PIPA531004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 452 and 693 of Human CPSF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 2, 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; Cleavage and polyadenylation specificity factor 100 kDa subunit; Cleavage and polyadenylation specificity factor subunit 2; CPSF 100 kDa subunit; CPSF 100kDa subunit; CPSF100 CPSF 100 kDa subunit CPSF 100kDa subunit cleavage and polyadenylation specificity factor 100 kDa subunit cleavage and polyadenylation specificity factor subunit 2
Gene Aliases: 100kDa; 2610024B04Rik; AI662483; Cpsf; CPSF100; CPSF2; KIAA1367; MCPSF; mKIAA1367
UniProt ID: (Human) Q9P2I0, (Mouse) O35218
Entrez Gene ID: (Human) 53981, (Mouse) 51786
Molecular Function: RNA binding protein endoribonuclease mRNA polyadenylation factor mRNA processing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.