Novus Biologicals
Manufacturer Code:NBP182910
Catalog # NBP182910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IELRKRVKLEGKELEEYLEKEKLKKEAAKKLEQSKEADIDSSDESDIEEDIDQPSAHKTKHDLMMKGEGSRKGSFFKQAKKSYPMFPAPEERIKWDEYGEIIKPEDFLVPELQATEEEKSKLESGLTNGDEPMDQDLSDVPTKCISTTESI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 2 100kDa Cleavage and polyadenylation specificity factor 100 kDa subunit cleavage and polyadenylation specificity factor subunit 2 CPSF 100 kDa subunit CPSF 100kDa subunit CPSF100 KIAA1367cleavage and polyadenylation specific factor 2 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; Cleavage and polyadenylation specificity factor 100 kDa subunit; Cleavage and polyadenylation specificity factor subunit 2; CPSF 100 kDa subunit; CPSF 100kDa subunit
Gene Aliases: CPSF100; CPSF2; KIAA1367
UniProt ID: (Human) Q9P2I0
Entrez Gene ID: (Human) 53981
Molecular Function:
RNA binding protein
endoribonuclease
mRNA polyadenylation factor
mRNA processing factor
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.