Novus Biologicals
Manufacturer Code:NBP257024
Catalog # NBP257024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AISLNITQKVHPVIWSLTSLPFDCTQALAVPKPIGGVVVFAVNSLLYLNQSVPPYGVALNSLTTGTTAFPLRT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 1 160kD subunit cleavage and polyadenylation specific factor 1 160kDa Cleavage and polyadenylation specificity factor 160 kDa subunit cleavage and polyadenylation specificity factor subunit 1 CPSF 160 kDa subunit CPSF160 HSU37012 P/cl.18 polyadenylation specificity factor; cleavage and polyadenylation specific factor 1, 160kDa; Cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; polyadenylation specificity factor
Gene Aliases: CPSF1; CPSF160; HSU37012; P/cl.18
UniProt ID: (Human) Q10570
Entrez Gene ID: (Human) 29894
Molecular Function: DNA binding protein RNA binding protein damaged DNA-binding protein mRNA polyadenylation factor mRNA processing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.