Novus Biologicals
Manufacturer Code:NB10079822
Catalog # NB10079822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1392 and 1442 of human cleavage and polyadenylation specific factor 1 160kD subunit using the numbering given in entry NP_037423.1 (GeneID 29894). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 1 160kD subunit cleavage and polyadenylation specific factor 1 160kDa Cleavage and polyadenylation specificity factor 160 kDa subunit cleavage and polyadenylation specificity factor subunit 1 CPSF 160 kDa subunit CPSF160 HSU37012 P/cl.18 polyadenylation specificity factor; cleavage and polyadenylation specific factor 1, 160kDa; Cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; polyadenylation specificity factor
Gene Aliases: CPSF1; CPSF160; HSU37012; P/cl.18
UniProt ID: (Human) Q10570
Entrez Gene ID: (Human) 29894
Molecular Function: DNA binding protein RNA binding protein damaged DNA-binding protein mRNA polyadenylation factor mRNA processing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.