Proteintech
Manufacturer Code:110311AP150UL
Catalog # 50-172-6812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPSF1 Fusion Protein Ag1489 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cleavage and polyadenylation specific factor 1, 160kDa; cleavage and polyadenylation specificity factor 1; Cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; CPSF 160 kDa subunit CPSF1 CPSF160 HSU37012 P/cl.18; CPSF160; polyadenylation specificity factor
Gene Aliases: CPSF1; CPSF160; HSU37012; P/cl.18
UniProt ID: (Human) Q10570, (Mouse) Q9EPU4
Entrez Gene ID: (Human) 29894, (Mouse) 94230, (Rat) 366952
Molecular Function:
DNA binding protein
RNA binding protein
damaged DNA-binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.