Proteintech
Manufacturer Code:122111AP150UL
Catalog # 50-172-7797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPOX Fusion Protein Ag2858 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: clone 560; coprogen oxidase; Coprogen oxidase Coproporphyrinogen oxidase Coproporphyrinogenase COX CPO CPOX CPX HCP; Coproporphyrinogen III oxidase, mitochondrial precursor (Coproporphyrinogenase) (Coprogen oxidase) (COX); coproporphyrinogen-III oxidase, mitochondrial; coproporphyrinogenase; COX; cpox {ECO:0000312|EMBL:AAI07665.1}; Oxygen-dependent coproporphyrinogen-III oxidase, mitochondrial
Gene Aliases: cac; CPO; CPOX; CPX; HCP; nct
UniProt ID: (Human) P36551, (Rat) Q3B7D0, (Mouse) P36552
Entrez Gene ID: (Human) 1371, (Rat) 304024, (Mouse) 12892
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.