Invitrogen
Manufacturer Code:PA571979
Catalog # PIPA571979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 138-166 amino acids from the N-terminal region of human CPNE6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: copine 6; Copine VI; copine VI (neuronal); Copine-6; Copine-6 Copine VI Neuronal-copine N-copine CPNE6; N-copine; neuronal copine; Neuronal-copine
Gene Aliases: AU067659; BB076446; CPNE6
UniProt ID: (Human) O95741, (Mouse) Q9Z140
Entrez Gene ID: (Human) 9362, (Mouse) 12891
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.