Proteintech
Manufacturer Code:137821AP150UL
Catalog # 50-172-9003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPNE6 Fusion Protein Ag4789 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: copine 6; Copine 6 Copine VI copine VI (neuronal) CPNE6 N COPINE Neuronal copine; Copine VI; copine VI (neuronal); Copine-6; N-copine; neuronal copine; Neuronal-copine
Gene Aliases: AU067659; BB076446; CPNE6
UniProt ID: (Human) O95741, (Mouse) Q9Z140
Entrez Gene ID: (Human) 9362, (Rat) 691478, (Mouse) 12891
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.