Invitrogen
Manufacturer Code:PA521876
Catalog # PIPA521876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 234 and 537 of Copine 3 (Uniprot IDNo.O75131) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Copine III; Copine-3; CPN3 PRO1071 copine-3
Gene Aliases: 5430428M23Rik; 5730450C07Rik; C85108; CPN3; CPNE3; KIAA0636; PRO1071
UniProt ID: (Human) O75131, (Mouse) Q8BT60
Entrez Gene ID: (Human) 8895, (Mouse) 70568
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.