Invitrogen
Manufacturer Code:PA541780
Catalog # PIPA541780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CPNE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromobindin 17; Copine I; Copine I CPN1 COPN1; copine III, like; Copine-1
Gene Aliases: 1810028N16Rik; COPN1; CPN1; CPNE1; cpne3; cpne3l; mKIAA4108; wu:fb69d07; wu:fc45a04; zgc:55873
UniProt ID: (Human) Q99829, (Bovine) Q08DB4, (Rat) D4A1R8, (Mouse) Q8C166
Entrez Gene ID: (Sheep) 100302101, (Dog) 477213, (Horse) 100054882, (Human) 8904, (Rabbit) 100328904, (Bovine) 615677, (Rat) 362249, (Mouse) 266692, (Zebrafish) 324841
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.