Invitrogen
Manufacturer Code:PA527412
Catalog # PIPA527412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 180 of Human Copine I |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromobindin 17; Copine I; Copine-1; COPN1 CPN1 copine-1
Gene Aliases: 1810028N16Rik; COPN1; CPN1; CPNE1; mKIAA4108
UniProt ID: (Human) Q99829, (Mouse) Q8C166
Entrez Gene ID: (Human) 8904, (Mouse) 266692
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.