Invitrogen
Manufacturer Code:PA519137
Catalog # PIPA519137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (APLKPLPPSAKDPA) corresponding to the C-terminus amino acids of CPNE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromobindin 17; Copine I; Copine-1; CPNE1 copine I COPN1 CPN1 MGC1142 OTTHUMP00000030775 OTTHUMP00000030776 copine-1
Gene Aliases: COPN1; CPN1; CPNE1
UniProt ID: (Human) Q99829
Entrez Gene ID: (Human) 8904
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.