Invitrogen
Manufacturer Code:PA543747
Catalog # PIPA543747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CPN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anaphylatoxin inactivator; Arginine carboxypeptidase; carboxypeptidase K; Carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; Carboxypeptidase N polypeptide 1 CPN FLJ40792 SCPN; Carboxypeptidase N small subunit; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; CPN; kininase I; Kininase-1; Lysine carboxypeptidase; Plasma carboxypeptidase B; SCPN; Serum carboxypeptidase N
Gene Aliases: 0610011F20Rik; ACBP; cb1037; CPN; CPN1; fa99g08; SCPN; wu:fa99g08; zgc:77485
UniProt ID: (Bovine) Q2KJ83, (Human) P15169, (Rat) Q9EQV8, (Mouse) Q9JJN5
Entrez Gene ID: (Horse) 100070412, (Rabbit) 100344758, (Bovine) 536753, (Dog) 477795, (Human) 1369, (Rat) 365466, (Guinea pig) 100725809, (Mouse) 93721, (Zebrafish) 337172
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.