Invitrogen
Manufacturer Code:PA514222
Catalog # PIPA514222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 52-81 amino acids from the N-terminal region of human CPN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anaphylatoxin inactivator; Arginine carboxypeptidase; carboxypeptidase K; Carboxypeptidase N catalytic chain; Carboxypeptidase N catalytic chain CPN Carboxypeptidase N polypeptide 1 Carboxypeptidase N small subunit Lysine carboxypeptidase Arginine carboxypeptidase Kininase-1 Serum carboxypeptidase N SCPN Anaphylatoxin inactivator Plasma carboxypeptidase B ACBP CPN1; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; Carboxypeptidase N small subunit; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; CPN; kininase I; Kininase-1; Lysine carboxypeptidase; Plasma carboxypeptidase B; SCPN; Serum carboxypeptidase N
Gene Aliases: 0610011F20Rik; ACBP; CPN; CPN1; SCPN
UniProt ID: (Human) P15169, (Mouse) Q9JJN5
Entrez Gene ID: (Human) 1369, (Mouse) 93721
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.